Anti-UBB Mouse Monoclonal Antibody (Unconjugated) [clone: 5B9-B3]
ROCK200-301-G61
- Antibody type:Primary
- Antigen name:Ubiquitin B
- Antigen symbol:UBB
- Clonality:Monoclonal
- Clone:5B9-B3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human,Bovine,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Gene ID:7314
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,250% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Ubiquitin Antibody was produced in mice by repeated immunizations raised against native bovine ubiquitin conjugated to KLH.
- Purification:Protein G purified
- Size:200 µg
- Pk:200 µG
Anti-Ubiquitin Antibody is recommended for use in WB and ELISA. Expect a band approximately ~10kDa corresponding to free ubiquitin. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:200; Western Blot: 1:1000
Type: Primary
Antigen: UBB
Clonality: Monoclonal
Clone: 5B9-B3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat, Bovine