Anti-Brain Natriuretic Peptide Mouse Monoclonal Antibody [clone: 27-4-3]
Catalog # USBIB2702-34B-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Brain Natriuretic Peptide
- Antigen symbol:BNP
- Clonality:Monoclonal
- Clone:27-4-3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized powder
- Amino acid number:1 to 21
- Storage buffer:200mM Na Citrate-Tris, 500mM sodium chloride, pH 7,5, 0,02% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human pro-BNP (aa 1-21) polylysinee conjugated (HPLGSPGSASDLETSGLQEQR)
- Purification:purified by Protein A chromatography
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: Brain Natriuretic Peptide
Clonality: Monoclonal
Clone: 27-4-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human,