Safety storage cabinets, type 90, S-PEGASUS-90
Supplier: asecos
Certificates
About this item
Fireproof safety cabinets, classified type 90 (fire resistant up to 90 minutes).
- Space saving folding doors
- Safety elements assembled outside the storage compartment for increased protection against corrosion
- Door open arrest system, convenient and safe removal or storage of containers
- Doors lockable with cylinder locking (integration in a locking system possible)
- Adjusting aids to compensate for uneven floor
- Integrated air ducts ready for connection (NW 75) to a technical exhaust system
Certifications: EN 14727 conformity (laboratory furniture).
GS approval, CE conformity.
Furnace tested (type test) in accordance with EN 14470-1
GS approval, CE conformity.
Furnace tested (type test) in accordance with EN 14470-1
Delivery: Supplied without interior equipment packages. Interior equipment packages need to be ordered in addition. Please contact the Avantor sales office for further information. Delivery prices available upon request.
Frequently Bought Together