Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-S100 Protein, beta Mouse Monoclonal Antibody [clone: 10K341]
  USBIS0052-11Q
Anti-S100 Protein, beta Mouse Monoclonal Antibody [clone: 10K341]
  USBIS0052-11Q

 

  • Antibody type:
    Primary
  • Antigen name:
    S100 Protein, beta
  • Clonality:
    Monoclonal
  • Clone:
    10K341
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    lyophilized powder
  • Storage buffer:
    1.2% sodium acetate, 2mg BSA, 0,01mg sodium azide. sterile ddH2O, PBS.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: Western Blot: 2?g/ml; Immunochemistry: 1-2?g/ml

Type: Primary
Antigen: S100 Protein, beta
Clonality: Monoclonal
Clone: 10K341
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat