Order Entry
Netherlands
ContactUsLinkComponent
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody [clone: 9A384]
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody [clone: 9A384]
Catalog # USBIP4378-56
Supplier:  US Biological
Anti-Polyoma Virus, Medium T Antigen Rat Monoclonal Antibody [clone: 9A384]
Catalog # USBIP4378-56
Supplier:  US Biological

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Polyoma Virus, Medium T Antigen
  • Clonality:
    Monoclonal
  • Clone:
    9A384
  • Conjugation:
    Unconjugated
  • Host:
    Rat
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    liquid
  • Storage buffer:
    PBS
  • Storage temperature:
    Cool
  • Concentration:
    ~1 mg/ml
  • Shipping temperature:
    Ice
  • Purification:
    purified
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Polyoma Virus, Medium T Antigen
Clonality: monoclonal
Clone: 9A384
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse