Anti-FceRI, alpha Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 10C204]
Catalog # USBIF0025-06B
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:FCERI, alpha
- Antigen symbol:FCERIA
- Clonality:Monoclonal
- Clone:10C204
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:Human
- Environmentally Preferable:
- Form:lyophilized powder
- Storage buffer:PBS, 1% BSA, 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 Tests
- Pk:100 Tests
Specifications
About this item
Do not freeze!
Type: Primary
Antigen: FceRI, alpha
Clonality: monoclonal
Clone: 10C204
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human