Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-Neuropeptide Y Y2 Receptor Rabbit Polyclonal Antibody
  USBI136934
Anti-Neuropeptide Y Y2 Receptor Rabbit Polyclonal Antibody
  USBI136934

 

  • Antibody type:
    Primary
  • Antigen name:
    Neuropeptide Y Y2 Receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Rat,
    Mouse
  • Environmentally Preferable:
  • Format:
    Serum
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µl
  • Pk:
    100 µl

 

 

Recommended dilution: Immunochemistry: 1:100-1:300

Type: Primary
Antigen: Neuropeptide Y Y2 Receptor
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat