Anti-CCNB2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI033382-APC
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:cyclin B2
- Antigen symbol:CCNB2
- Clonality:Polyclonal
- Conjugation:APC (Allophycocyanin)
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Form:liquid
- Modification:phospho-Ser10
- Storage buffer:PBS, pH 7,2, contains ~0,09% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: FLISA: 1:1000; Dot blot 1:500
Do not freeze!
Type: Primary
Antigen: CCNB2 (cyclin B2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human