Order Entry
Netherlands
ContactUsLinkComponent
Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # ROCK700-105-096
Anti-FITC Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # ROCK700-105-096

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Fluorescein
  • Antigen symbol:
    FITC
  • Clonality:
    Polyclonal
  • Conjugation:
    AP (Alkaline Phosphatase)
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
  • Reactivity:
    Multiple
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    none
  • Format:
    Antibody fraction
  • Form:
    Liquid (sterile filtered)
  • Antibody fragment:
    F(ab')2
  • Storage buffer:
    0,05 M Tris Chloride, 0,15M Sodium Chloride, 0,001M Magnesium Chloride, 0,0001M Zinc Chloride, 50% (v/v) Glycerol
  • Storage temperature:
    Cool
  • Concentration:
    1,0 mg/ml by UV absorbance at 280 nm
  • Shipping temperature:
    Ice
  • Immunogen:
    Fluorescein conjugated to Goat IgG
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    1 mg
  • Pk:
    1 mg

Specifications

About this item

Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:38000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: pan