Anti-His-Tag Rabbit Polyclonal Antibody (Biotin)
GENSA00205-40
- Antibody type:Tag
- Antigen name:His-tag
- Antigen symbol:His
- Clonality:Polyclonal
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:40 µg
- Pk:40 µG
Type: Tag
Antigen: His-Tag (His-Tag)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: