Anti-MYC Mouse Monoclonal Antibody [clone: 9E11]
The c-Myc Antibody (9E11) from Novus Biologicals is a mouse monoclonal antibody to c-Myc. This antibody reacts with human, mouse, chicken, yeast. The c-Myc Antibody (9E11) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: 9E11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Mouse, Chicken, Yeast
- Catalog No:
- NOVUNB200-108
- NOVUNB200-108SS
- Antigen symbol:
- c-Myc
- c-Myc
- Antigen name:
- v-myc avian myelocytomatosis viral oncogene homolog
- v-myc avian myelocytomatosis viral oncogene homolog
- Antigen synonyms:
- MRTL|bHLHe39|c-Myc|MYCC
- MRTL|bHLHe39|c-Myc|MYCC
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 9E11
- 9E11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 4609
- 4609
- Isotype:
- IgG2a kappa
- IgG2a kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- ChIP:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Frequently Bought Together