Anti-TGF beta Mouse Monoclonal Antibody
Catalog # BIRBORB76826-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:TGF beta
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Format:whole IgG
- Storage buffer:0,01M PBS, pH 7.0
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:TGF-beta from human platelets
- Purification:purified by Protein G chromatography
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Immunochemistry: 1:1000; Immunochemistry: 1:1000
Type: Primary
Antigen: TGF beta
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human