Bestelling invoeren
Netherlands
ContactUsLinkComponent
Anti-O GlcNAc transferase Rabbit Polyclonal Antibody
Anti-O GlcNAc transferase Rabbit Polyclonal Antibody
Leverancier:  Biorbyt
Certificaten
Anti-O GlcNAc transferase Rabbit Polyclonal Antibody
Leverancier:  Biorbyt

Specificaties

  • Catalog No:
  • BIRBORB1121-100
  • BIRBORB1121-200
  • Type antilichaam:
  • Primair
  • Primair
  • Clonality:
  • Polyklonaal
  • Polyklonaal
  • Conjugatie:
  • Onconjugeerd
  • Onconjugeerd
  • Host:
  • Rabbit
  • Rabbit
  • Isotype:
  • IgG
  • IgG
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemie:
  • Yes
  • Yes
  • Immunogeen:
  • Synthetic peptide
  • Synthetic peptide
  • Formulier:
  • liquid
  • liquid
  • Purification:
  • purified by Protein A chromatography
  • purified by Protein A chromatography
  • Concentratie:
  • 0,5 mg/ml
  • 0,5 mg/ml
  • Opslagbuffer:
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Opslagtemperatuur:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specificaties

Gerelateerde informatie

Frequently Bought Together