Order Entry
Netherlands
ContactUsLinkComponent
Anti-CAMKK1 Mouse Monoclonal Antibody [clone: 4B4]
Anti-CAMKK1 Mouse Monoclonal Antibody [clone: 4B4]
Catalog # ABNOH00084254-M02
Supplier:  Abnova
undefined
Anti-CAMKK1 Mouse Monoclonal Antibody [clone: 4B4]
Catalog # ABNOH00084254-M02
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium/Calmodulin-Dependent Protein Kinase Kinase 1, alpha
  • Antigen symbol:
    CAMKK1
  • Clonality:
    Monoclonal
  • Clone:
    4B4
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    84254
  • Antigen synonyms:
    MGC34095|Ca2+/calmodulin-dependent protein kinase kinase alpha|CaM-kinase kinase 1|CaMKK 1|Calcium/calmodulin-dependent protein kinase kinase alpha|Calcium/calmodulin dependent protein kinase kinase alpha|CaMKK|CaMKK alpha|Calcium/calmodulin dependent protein kinase kinase 1 alpha|DKFZp761M0423|CaM-kinase IV kinase|Camkk1|Calcium/calmodulin dependent protein kinase kinase|CAMKK alpha protein|KKCC1_HUMAN|CaM-KK alpha|CAMKKA|Calcium/calmodulin-dependent protein kinase kinase 1|CaM-kinase kinase alpha|CaM-KK 1|CaM kinase kinase alpha|OTTMUSP00000006419.|Calcium/calmodulin dependent protein kinase kinase 1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

Specifications

About this item

Type: Primary
Antigen: CAMKK1
Clonality: Monoclonal
Clone: 4B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human