Anti-TGIF2 Mouse Monoclonal Antibody [clone: 2D4]
Catalog # ABNOH00060436-M16
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:TGFB-induced factor homeobox 2
- Antigen symbol:TGIF2
- Clonality:Monoclonal
- Clone:2D4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:60436
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: TGIF2
Clonality: Monoclonal
Clone: 2D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human