Anti-ACSS2 Mouse Monoclonal Antibody [clone: 2A10]
Catalog # ABNOH00055902-M09
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:acyl-CoA synthetase short-chain family member 2
- Antigen symbol:ACSS2
- Clonality:Monoclonal
- Clone:2A10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:55902
- Antigen synonyms:ACS|ACECS|ACAS2|dJ1161H23.1|ACSA
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ACSS2
Clonality: Monoclonal
Clone: 2A10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human