Anti-RUVBL2 Mouse Polyclonal Antibody
Catalog # ABNOH00010856-B01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:RuvB-like 2
- Antigen symbol:RUVBL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:10856
- Antigen synonyms:REPTIN|RVB2|TAP54-beta|INO80J|ECP-51|RuvB like AAA ATPase 2|TIP48|TIH2|CGI-46|TIP49B|ECP51
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µg
Specifications
About this item
Type: Primary
Antigen: RUVBL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human