Anti-MAP2K5 Mouse Monoclonal Antibody [clone: M1-E6]
Catalog # ABNOH00005607-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase Kinase 5
- Antigen symbol:MAP2K5
- Clonality:Monoclonal
- Clone:M1-E6
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:5607
- Antigen synonyms:PRKMK5|MEK5|MAPKK5|HsT17454
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: MAP2K5
Clonality: Monoclonal
Clone: M1-E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human