Anti-DAO Mouse Monoclonal Antibody [clone: 2F5]
Catalog # ABNOH00001610-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:D-amino-acid oxidase
- Antigen symbol:DAO
- Clonality:Monoclonal
- Clone:2F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:1610
- Antigen synonyms:DAAO|DAO1|DAMOX
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: DAO
Clonality: Monoclonal
Clone: 2F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human