Order Entry
Netherlands
ContactUsLinkComponent
 
Anti-CRYGD Mouse Monoclonal Antibody [clone: 4E12]
  ABNOH00001421-M03
 :  Abnova
undefined
Anti-CRYGD Mouse Monoclonal Antibody [clone: 4E12]
  ABNOH00001421-M03
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Crystallin, gamma D
  • Antigen symbol:
    CRYGD
  • Clonality:
    Monoclonal
  • Clone:
    4E12
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Gene ID:
    1421
  • Antigen synonyms:
    cry-g-D|CCA3|CCP|CTRCT4|CRYG4|PCC|CACA
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µg

 

 

Type: Primary
Antigen: CRYGD
Clonality: Monoclonal
Clone: 4E12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human