Anti-ATP6V1A Mouse Monoclonal Antibody [clone: 4F5]
Catalog # ABNOH00000523-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:ATPase, H+ transporting, lysosomal 70kDa, V1 subunit A
- Antigen symbol:ATP6V1A
- Clonality:Monoclonal
- Clone:4F5
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Gene ID:523
- Antigen synonyms:VPP2|Vma1|VA68|ATP6A1|ATP6V1A1|HO68
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: ATP6V1A
Clonality: Monoclonal
Clone: 4F5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human