Anti-YY1 Rabbit Polyclonal Antibody
Catalog # ABGEAP2517A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:YY1 transcription Factor
- Antigen symbol:YY1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-Terminal ( 75–125 aa)
- Cross adsorption:No
- Antigen synonyms:delta transcription factor|YIN-YANG-1|UCRBP|INO80S|transcriptional repressor protein YY1|Yin and Yang 1 protein|YY-1|DELTA|NF-E1|INO80 complex subunit S
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: YY1 (YY1 transcription factor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human