Anti-SPOCK2 Rabbit Polyclonal Antibody
AVIVARP62235_P050
- Antibody type:Primary
- Antigen name:sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2
- Antigen symbol:SPOCK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9806
- Antigen synonyms:testican-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SPOCK2 (sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: