Anti-ENOPH1 Rabbit Polyclonal Antibody
AVIVARP57530_P050
- Antibody type:Primary
- Antigen name:enolase-phosphatase 1
- Antigen symbol:ENOPH1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:middle
- Cross adsorption:none
- Gene ID:58478
- Antigen synonyms:MST145|E1|mtnC|MASA
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: ENOPH1 (enolase-phosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: