Order Entry
Netherlands
ContactUsLinkComponent
Waste disposal boxes MultiSafe opti
Waste disposal boxes MultiSafe opti
Catalog # SEMA5724
Supplier:  SEMADENI
Waste disposal boxes MultiSafe opti
Catalog # SEMA5724
Supplier:  SEMADENI
Specifications
  • Ø×H:
    97x164 mm
  • Capacity:
    700 ml
  • Description:
    MultiSafe opti 700
  • Seq:
    01
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Boxes for the disposal of scalpels, cannulas, disposable syringes, tubing, broken glass and long-stemmed objects (e.g. pipettes).
  • Round container form
  • Hinged day lid
  • The size of the disposal opening can be further enlarged by breaking out section s as required
  • A variety of holders are available to fasten the boxes securely to walls or surfaces so they won`t overturn
  • Lateral grooves on the lid confirm closure visually