Order Entry
Netherlands
ContactUsLinkComponent
Active pirani gauges
Active pirani gauges
Certificates
Active pirani gauges
Specifications
  • Catalog No:
  • 619-0221
  • 619-0220
  • Model:
  • APG-L-NW16
  • APG-M-NW16
  • Accuracy:
  • 1×10ˉ4 mbar
  • 1×10ˉ3 mbar
  • Permitted ambient temperature for operation:
  • +10...+60 °C
  • +10...+60 °C
  • Permitted ambient temperature for storage:
  • 0...+70 °C
  • 0...+70 °C
  • Power input:
  • 20 to 36 V dc (max 1 V ripple)
  • 20 to 36 V dc (max 1 V ripple)