PCR tube racks with lid, 96-well
Supplier: DELTALAB
Certificates
About this item
Racks made of PP. Suitable for PCR individual tubes, 96 well plate and strips of 8 or 12 tubes.
- Alphanumerical grid helps in sample identification
- Complete with transparent lid
- It can be used alone or inserted with workstation
- Withstands temperature from −90 to 121 °C
Lid has a non-slip surface to allow safe storage of multiple racks.
W×D×H: 125×85×15 mm
Frequently Bought Together