Tough-Spots® Label, Rolls
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
These Tough-Spots® PVC label rolls are available in a variety of sizes, colors and formats.
- Temperature resistant from −196 to +80 °C
- Accept writing from most lab markers
The dispensing box assists in labeling individual tubes used in small experiments.
Frequently Bought Together