Multi-channel pipettes, electronic, variable volume, Ovation BioNatural™
Supplier: VISTALAB TECHNOLOGIES
Certificates
About this item
This pipette is an adjustable volume, air displacement, fully electronic motor-driven pipette intended to aspirate and dispense precise fluid volumes. 8 and 12-channel models, in both left and right-handed configurations, are available to cover liquid dispensing needs between 0,5 and 1250 µl. Ideal for handling repetitive and complex liquid handling tasks.
- Ergonomic design reduces fatigue and repetitive stress injuries
- Larger grip surface for more comfort during operation
- Easy tip ejection with minimal thumb force
- LCD indicates liquid handling functions, status, settings, and remaining battery power
- Keypad is easy to use and provides access to all liquid handling functions and settings
Frequently Bought Together