Gas cartridge filters, Super Clean™
Supplier: Thermo Fisher Scientific
Certificates
About this item
These filters can be easily replaced without tools.
- 99,9999% pure gas (or 6,0 grade) output
- No carrier gas line contamination during cartridge change
- Maximum pressure of 11 bar (160 psi)
- Maximum flow of 7 L/min
Certifications: TUEV approved.
Cartridge filter baseplates/manifolds
Product Information
Frequently Bought Together