(2-Carboxyethyl)triphenylphosphonium chloride ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:36626-29-6
:
Formula:C₂₁H₂₀ClO₂P
MDL Number:MFCD00185663
Molecular Weight:370.81 g/mol
Melting Pt:197…201 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together