Order Entry
Netherlands
ContactUsLinkComponent
High MW genomic DNA isolation kits, MegaLong™
High MW genomic DNA isolation kits, MegaLong™
Supplier:  G-Biosciences
Certificates
undefined
High MW genomic DNA isolation kits, MegaLong™
Supplier:  G-Biosciences

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Related Information

Frequently Bought Together