ContactUsLinkComponent
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hosebarb, 1/8" ID; 25/PK
  MFLX45518-11
CE certificate
undefined
Masterflex® Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hosebarb, 1/8" ID; 25/PK
  MFLX45518-11
CE certificate
  • Model:
  • 45518-11
  • 45518-24
  • 30800-16
  • 45518-26
  • 45518-13
  • 45518-08
  • 30800-26
  • 45518-02
  • 30800-28
  • 45518-06
  • 30800-22
  • 30800-24
  • 45518-00
  • 30800-18
  • 45518-44
  • 30800-20
  • 45518-22
  • Connection type (port 1):
  • Male luer lock
  • Male luer slip
  • Male luer lock
  • Male luer slip
  • Male luer lock
  • Male luer lock
  • Male luer slip
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer lock
  • Male luer slip
  • Male luer slip
  • Male luer slip
  • Connection type (port 2):
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Barb
  • Hose barb
  • Hose Barb
  • Hose barb
  • Hose barb
  • Hose barb
  • Hose barb
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  • 3 Months
  •  
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilisation.
  • Barb size/tubing int.Ø (port 1):
    1/8"
  • Connection Type:
    Male Luer to Barb
  • Flow pattern:
    Straight
  • Description:
    Fitting, Polypropylene, Straight, Male Luer Lock to Low-Profile Hosebarb, 1/8" ID; 25/PK
  • Seq:
    13
  • Environmentally Preferable:
  • Model:
    45518-11
  • Connection type (port 1):
    Male luer lock
  • Connection type (port 2):
    Hose barb
  • No. of ports:
    2
  • Pk:
    25
  • Fitting material:
    Polypropylene
Frequently Bought Together