Zinc standard solution, concentrate, 1000 mg Zn in 0,06% hydrochloric acid (from ZnCl₂) Titrisol® standard for AAS, Supelco®
1.09953.0001
: Merck
:
- Concentration:1000 mg
- Matrix:2 M HNO₃
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- Environmentally Preferable:
- M-Clarity quality level:MQ100
- Element:Zinc
- Symbol:Zn
: Traceable to SRM from NIST