Palladium 10% on activated carbon hydrogenation catalyst (oxidised form), for synthesis, unreduced, Sigma-Aldrich®
: Merck
:7440-05-3
:
:Pd
:MFCD00011167
:106.420 g/mol
:M-Clarity™ quality level = MQ200
:231-115-6
- Environmentally Preferable:
- Solubility:(20 °C) insoluble
- Bulk Density:400 kg/m3
- pH:6 - 7 (50 g/l, H2O, 20 °C) (slurry)
- Ignition Point:460 °C
Frequently Bought Together