Water, LiChrosolv® for chromatography, for LC-MS, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Hydrogen oxide
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
Boiling Pt:100
Melting Pt:0
Density:1 g/cm³ (4 °C)
Molecular Weight:18.02 g/mol
Merck Index:13,10098
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:231-791-2
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Related Information
Certificates
Search by Lot NumberMERC1.15333.1000_TDS_EN MERC1.15333.2500_TDS_EN ProductPage_115333
Product specification
Merck Pharma Segment Brochure
Product Literature
msds_115333
SDS
Frequently Bought Together