ContactUsLinkComponent
Fraction collector, Frac-920
Catalog # 18-1177-40
Supplier:  Cytiva
Fraction collector, Frac-920
Catalog # 18-1177-40
Supplier:  Cytiva
Specifications
  • Capacity:
    175 fractions in 12-mm Ø tubes; 95 fractions in 10-18 mm Ø tubes; 40 fractions in 30-mm Ø tubes
  • Description:
    Fraction collector, Frac-920
  • Seq:
    01
  • Environmentally Preferable:
  • Fraction timer:
    0,01 - 9,999 min
  • Fraction volume timer:
    0,1 - 999 ml
  • Pk:
    1
Specifications
About this item
Frac-920 is an automated fraction collector for robust and reliable collection of fractions based on time or volume. Frac-920 can be used with ÄKTAdesign chromatography system or as a stand-alone instrument. It has a carousel design and a small footprint.
  • For standalone use, as well as use with UNICORN control software
  • Robust and simple to use
  • Easy and fast method programming
Fraction collector Frac-920 can collect up to 175 fractions in 12-mm diameter tubes, up to 95 fractions in 10-18 mm diameter tubes, and up to 40 fractions in 30-mm diameter tubes.