ContactUsLinkComponent
Homogenisers, Polytron® PT 1200 E and PT 1300 D
undefined
Homogenisers, Polytron® PT 1200 E and PT 1300 D
  • Catalog No:
  • 431-0136
  • 431-0138
  • Model:
  • Polytron® PT 1300 D (digital)
  • Polytron® PT 1200 E (Ecoline)
  • Volume H₂O:
  • 0,1 - 250 ml
  • 0,1 - 250 ml
  • Speed range:
  • 7000 - 30000 min⁻¹
  • 0 - 25000 min⁻¹
  • Speed display:
  • Digital
  • No
  • Motor rating:
  • 100 W
  • 100 W
  • Weight:
  • 0,6 kg
  • 0.44
  • W×D×H:
  • 40×45×210 mm
  • 44x55x220
Frequently Bought Together