Laboratory trays, LaboPlast®
About this item
PP, white.
- Manufactured using the injection moulding process, not by deep drawing
- Trays can be nested
- Trays not suitable for temperature controlled cupboards or dishwashers
- Temperature resistent up to 60 °C (briefly up to 80 °C)
- With EU foodstuffs approval
The laboratory trays are practical all-purpose trays that can be nested when empty, also durable, shatterproof and resist flexing. The trays have a strong, rounded rim. Suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as an instrument holder, a medicine tray, a photographic development basin, a goods presentation tray and a display tray for foodstuffs.
8 Options Available Below
Product Details & Documents
PP, white.
- Manufactured using the injection moulding process, not by deep drawing
- Trays can be nested
- Trays not suitable for temperature controlled cupboards or dishwashers
- Temperature resistent up to 60 °C (briefly up to 80 °C)
- With EU foodstuffs approval
The laboratory trays are practical all-purpose trays that can be nested when empty, also durable, shatterproof and resist flexing. The trays have a strong, rounded rim. Suitable as a spill trough for environmentally dangerous, aggressive substances, for experiments, as an etching tray, as an instrument holder, a medicine tray, a photographic development basin, a goods presentation tray and a display tray for foodstuffs.
Documents
Certificates
Search by Lot NumberProduct Family Options
Trays
Product Information
Tray sets
Product Information
- CapacityBaseW×D×HAvailabilityPrice
- 216-0899Laboratory tray set of 4 with one tray each 0,5/1,5/3,0/10,0 lLaboratory tray set of 4 with one tray each 0,5/1,5/3,0/10,0 l--- mmLog in for availability1 * 4 items£70.60
- 216-0922Laboratory tray set of 6 with one tray each 0,5/1,5/3,0/10,0/21,0/39,0 lLaboratory tray set of 6 with one tray each 0,5/1,5/3,0/10,0/21,0/39,0 l--- mmLog in for availability1 * 1 SET£203.00