Bottles, narrow neck, round, HDPE, low particulate, IP2, with screw caps, Nalgene®
Supplier: Thermo Fisher Scientific
Certificates
About this item
HDPE, translucent with PP screw cap.
- Good chemical resistance
- Screw cap supplied assembled on the bottle
- Leakproof
Bottles with an average of less than 30 particles per ml at 0,3 µm and greater. Excellent for storing high-purity chemicals. Product includes a certificate of quality that assures the product has been tested and accepted in accordance with Nalgene® specifications. Low particulate IP 2 bottles can be used in compliance with the regulations for customers who are designing, assembling and certifying their own combination packaging.
Specifications
- Catalog No:
- 215-1279
- 215-1272
- 215-1271
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement