Indium tin alloy (52:48 w%), eutectic ≥99.99% (metals basis), ingot
Supplier: Thermo Fisher Scientific
Certificates
About this item
:
MDL Number:MFCD00144477
Melting Pt:117 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together