Order Entry
Northern Ireland
ContactUsLinkComponent
 
Di-tert-butyl peroxide 99%
 
undefined
Di-tert-butyl peroxide 99%

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3880 to 1.3900 (20°C, 589 nm)
  • GC:
    ≥98.5 %

 

 

Frequently Bought Together