Rubbermaid® Trademaster® Carts
The TradeMaster® cart with four drawers is a complete tool storage and mobile workbench system, with shelving and cabinet configurations to meet any storage needs.
- Metal-reinforced structural foam shelves engineered for durability
- Moulded in storage compartments for small parts or tools
- Supports up to 340 kg (with 129-1827)
Two non-marking 12,7 cm casters and two swivel casters with full-locking mechanism.
Frequently Bought Together