Long Fragment Buffer Expansion
Catalog # OXNTEXP-LFB001
Supplier: Oxford Nanopore Technologies
CAS Number:
Specifications
- Pk:1 KIT
- Application:
- Description:LFB expansion
- Environmentally Preferable:
- Compatible with:All flow cells
- No. of reactions:4
Specifications
About this item
Additional Long Fragment Buffer (LFB).
The LFB expansion contains additional Long Fragment Buffer (LFB) for use with sequencing experiments where shorter fragments (<3 kb) are to be removed. The LFB expansion contains enough reagents for four reactions.
Delivery: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at −20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.