ContactUsLinkComponent
Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
undefined
Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
  • Model:
  • 30401-58
  • 31702-61
  • 30401-60
  • 31702-57
  • 30401-55
  • 30401-59
  • 30401-56
  • 31208-48
  • 30401-54
  • 31208-46
  • 31208-47
  • 31208-44
  • 31208-45
  • 31208-41
  • 30401-63
  • 31702-47
  • 31702-45
  • 31702-62
  • 31208-42
  • Orifice dimensions:
  • 0,34375"
  • 0,25"
  • 0,1875"
  • 0,1875"
  • 0,09375"
  • 0,09375"
  • 0,09375"
  • 0,0625"
  • 0,1875"
  • 0,0625"
  • 0,0625"
  • 15,08 mm (0,59375")
  • 11,9 mm (0,46875")
  • 0,34375"
  • 0,0625"
  • Sterilisation:
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Autoclave, ethylene oxide (ETO), or gamma
  • Ethylene oxide (ETO) only
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (EtO)
  • Ethylene oxide (ETO) only
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Autoclave or ethylene oxide (ETO)
  • Ethylene oxide (ETO) only
  • Autoclave, ethylene oxide (ETO), or gamma
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 1 year
Frequently Bought Together