Potassium permanganate 0,02 mol/l 0,1 N in aqueous solution, AVS TITRINORM volumetric solution standardised against ferrous ethylenediammonium sulphate
Catalog # 31456.295
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2% accuracy):0.01996 - 0.02004 mol/L
- NIST traceable:Confirmed
- Conforms to Reag.Ph.Eur.:Passes test
- Standardised against C₂H₈N₂.Fe.2H₂SO₄:Passes test
Specification Test Results
About this item
Synonyms: Permanganic acid potassium salt
:
:158.03 g/mol
UN:3082
:GHS09
CAS Number:7722-64-7
Formula:KMnO₄
MDL Number:MFCD00011364
ADR:9 III
Merck Index:12,07824