VWR®, Launderable cleanroom socks
Cleanroom socks provide comfort and durability for limited and extended wear in cleanroom environments.
- 100% polyester
- Colour coded bands for visual identification
- Cleanroom compatible
- No particle generation
- Launderable
Minimise the risk of particle contamination with VWR® cleanroom socks.
Ideal for use with VWR® cleanroom clogs.
These launderable cleanroom socks provide comfort for extended wear and are cleanroom compatible.
Independent tests recommend that VWR launderable socks can withstand a minimum of 60 laundering cycles with less than 10% shrinkage.
: Socks are packaged 1 pair per bag, 200 bags per case.
- Catalog No:
- 113-9939
- 113-9937
- 113-9936
- 113-9938
- Garment application:
- Cleanroom
- Cleanroom
- Cleanroom
- Cleanroom
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Polyester
- Polyester
- Polyester
- Polyester
Frequently Bought Together