Chloramine-T (sodium salt) trihydrate, extra pure, Supelco®
: Merck
: Tosylchloramide sodium trihydrate, Chlorine bleaching solution, Chloramine T trihydrate, Chloramine T sodium salt trihydrate, Sodium chloro(tosyl)amide trihydrate, N-Chloro-4-toluenesulphonamide sodium salt
:7080-50-4
:
:CH₃C₆H₄SO₂NClNa·3H₂O
:MFCD00149066
:1.43 g/cm³ (20 °C)
:281.69 g/mol
:167…170 °C
:192 °C (closed cup)
:6.1 (8) II
:13,02085
:M-Clarity™ quality level = MQ200
:204-854-7
:2928
:GHS05
:GHS08
:GHS07
:Refrigerator
- Environmentally Preferable:
- Assay (iodometric):98.0 - 103.0 %
- Identity (IR-spectrum):passes test
- Appearance of solution:passes test
- pH-value (5 %; water):8.0 - 10.0
- Matter insoluble in ethanol:≤ 2 %
- ortho compounds:passes test
- Residual solvents (ICH Q3C):excluded by manufactoring process
Frequently Bought Together