ContactUsLinkComponent
Micro spatula, stainless steel
Micro spatula, stainless steel
Catalog # BURK5386-0001
Supplier:  BURKLE
undefined
Micro spatula, stainless steel
Catalog # BURK5386-0001
Supplier:  BURKLE
Specifications
  • Capacity:
    1,25 ml
  • Length:
    170 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The micro spatula can be used for reliably measuring, sampling, or processing small amounts of powder, chemicals, pastes, creams, or active ingredients.
  • Stainless steel AISI 304 (1.4301)
  • High-gloss polished
The laboratory scoop is manufactured entirely without edges, crevices, or undercuts. This eliminates the possibility of impurities or cross-contamination, as no product residue or cleaning agents can be deposited. The micro spatula is ideal for use in the pharmaceutical industry as well as food and beverage, and cosmetics.