Order Entry
Northern Ireland
ContactUsLinkComponent
5(6)-TAMRA free acid ≥97% (by HPLC) amine-reactive fluorescent dye
5(6)-TAMRA free acid ≥97% (by HPLC) amine-reactive fluorescent dye
Catalog # 90027.
Supplier:  Biotium
CAS Number:  
undefined
5(6)-TAMRA free acid ≥97% (by HPLC) amine-reactive fluorescent dye
Catalog # 90027.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440
  • Environmentally Preferable:

Specifications

About this item

Synonyms: TAMRA 5 and 6-isomer, Tetramethylrhodamine-5-(and-6)-maleimide, 5(6)-Carboxy-tetramethylrhodamine, 9-[2,4(or 2,5)-Dicarboxyphenyl]-3,6-bis(dimethylamino)-xanthylium inner salt
 :
 :430.45 g/mol
Storage Temperature:Freezer
CAS Number:98181-63-6
 :MFCD00467499
Formula:C₂₅H₂₂N₂O₅
MDL Number:MFCD16661194